Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332470.1 | NP_001321244.1 | 1 - 2 | 359 | 63 | |
NM_101936.3 | NP_173507.1 | 2 - 3 | 373 | 63 | ||
GenBank mRNA | AK118972.1 | - | 2 - 3 | 349 | 63 | |
BT006034.1 | - | 2 - 3 | 336 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |