Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STL2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126208.4 | NP_178256.1 | 2 - 3 | 417 | 68 | |
GenBank mRNA | AY074332.1 | - | 2 - 3 | 408 | 68 | |
AY123030.1 | - | 2 - 3 | 379 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |