Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126290.2 | NP_178338.1 | 1 - 2 | 382 | 67 | |
GenBank mRNA | AY649301.1 | - | 1 - 2 | 359 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |