Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111012.4 | NP_186795.1 | 5 - 6 | 3424 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |