Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC48C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337349.1 | NP_001326134.1 | 7 - 8 | 2344 | 121 | |
NM_111027.4 | NP_186810.2 | 7 - 8 | 2385 | 121 | ||
GenBank mRNA | AX765816.1 | - | 7 - 8 | 2208 | 121 | |
AY136443.1 | - | 7 - 8 | 2241 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |