Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111071.3 | NP_186854.1 | 3 - 4 | 734 | 81 | |
GenBank mRNA | AY062747.1 | - | 3 - 4 | 731 | 81 | |
BT003375.1 | - | 3 - 4 | 532 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |