Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPBETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111075.3 | NP_186858.1 | 7 - 8 | 1487 | 75 | |
NM_180169.1 | NP_850500.1 | 7 - 8 | 1475 | 75 | ||
GenBank mRNA | BT000662.1 | - | 7 - 8 | 1463 | 75 | |
BT001915.1 | - | 7 - 8 | 1400 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |