Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125558.1 | NP_001119030.1 | 10 - 11 | 1609 | 60 | |
NM_118334.5 | NP_193944.2 | 14 - 15 | 2302 | 60 | ||
GenBank mRNA | AY518293.1 | - | 14 - 15 | 2065 | 60 | |
FJ708751.1 | - | 14 - 15 | 1957 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |