Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOBER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118356.4 | NP_193961.3 | 3 - 4 | 522 | 109 | |
GenBank mRNA | AK117563.1 | - | 3 - 4 | 471 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |