Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084964.1 | NP_001078433.1 | 2 - 3 | 423 | 118 | |
NM_001203877.1 | NP_001190806.1 | 2 - 3 | 524 | 118 | ||
NM_118430.5 | NP_194032.2 | 2 - 3 | 503 | 118 | ||
GenBank mRNA | AK227353.1 | - | 2 - 3 | 336 | 118 | |
BT024728.1 | - | 2 - 3 | 152 | 118 | ||
DQ015901.1 | - | 2 - 3 | 152 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |