Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118480.4 | NP_567685.1 | 1 - 2 | 600 | 69 | |
GenBank mRNA | AY084337.1 | - | 1 - 2 | 336 | 69 | |
BT003955.1 | - | 1 - 2 | 341 | 69 | ||
BT005054.1 | - | 1 - 2 | 227 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |