Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GA2OX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102743.3 | NP_174296.1 | 2 - 3 | 1059 | 76 | |
GenBank mRNA | AJ132436.1 | - | 2 - 3 | 841 | 76 | |
BT002987.1 | - | 2 - 3 | 868 | 76 | ||
BT004464.1 | - | 2 - 3 | 747 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |