Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332883.1 | NP_001320726.1 | 10 - 11 | 1484 | 113 | |
NM_001332884.1 | NP_001320727.1 | 10 - 11 | 1620 | 113 | ||
NM_102766.6 | NP_564353.1 | 10 - 11 | 1538 | 113 | ||
GenBank mRNA | AY035226.1 | - | 10 - 11 | 1068 | 113 | |
AY056419.1 | - | 10 - 11 | 1440 | 113 | ||
AY090322.1 | - | 10 - 11 | 1068 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |