Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332971.1 | NP_001320846.1 | 4 - 5 | 1229 | 81 | |
NM_102900.5 | NP_174446.2 | 6 - 7 | 1430 | 81 | ||
GenBank mRNA | AK222010.1 | - | 6 - 7 | 1377 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |