Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP97A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102914.3 | NP_564384.1 | 4 - 5 | 673 | 73 | |
GenBank mRNA | AY056446.1 | - | 4 - 5 | 668 | 73 | |
AY058173.1 | - | 4 - 5 | 673 | 73 | ||
AY142017.1 | - | 4 - 5 | 635 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |