Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102953.4 | NP_174499.1 | 10 - 11 | 1303 | 115 | |
NM_179407.3 | NP_849738.1 | 10 - 11 | 1303 | 115 | ||
GenBank mRNA | AY093169.1 | - | 10 - 11 | 1188 | 115 | |
BT008758.1 | - | 10 - 11 | 1130 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |