Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204036.1 | NP_001190965.1 | 4 - 5 | 991 | 119 | |
NM_001342538.1 | NP_001320167.1 | 4 - 5 | 888 | 119 | ||
NM_120097.3 | NP_568058.1 | 4 - 5 | 922 | 119 | ||
GenBank mRNA | AF302675.1 | - | 4 - 5 | 752 | 119 | |
AF370208.1 | - | 4 - 5 | 887 | 119 | ||
AY133770.1 | - | 4 - 5 | 734 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |