Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337082.1 | NP_001324514.1 | 12 - 13 | 985 | 96 | |
NM_001337083.1 | NP_001324515.1 | 12 - 13 | 893 | 96 | ||
NM_129998.3 | NP_566016.1 | 12 - 13 | 934 | 96 | ||
NM_180084.2 | NP_850415.1 | 12 - 13 | 937 | 96 | ||
GenBank mRNA | AF387018.1 | - | 12 - 13 | 936 | 96 | |
BT006613.1 | - | 11 - 12 | 802 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |