Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337082.1 | NP_001324514.1 | 8 - 9 | 650 | 74 | |
NM_001337083.1 | NP_001324515.1 | 8 - 9 | 558 | 74 | ||
NM_129998.3 | NP_566016.1 | 8 - 9 | 599 | 74 | ||
NM_180084.2 | NP_850415.1 | 8 - 9 | 602 | 74 | ||
GenBank mRNA | AF387018.1 | - | 8 - 9 | 601 | 74 | |
BT006613.1 | - | 7 - 8 | 467 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |