Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPSAb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111349.4 | NP_187128.1 | 1 - 2 | 426 | 86 | |
NM_180184.2 | NP_850515.1 | 1 - 2 | 184 | 86 | ||
GenBank mRNA | U66223.1 | - | 1 - 2 | 155 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |