Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120181.5 | NP_195724.1 | 1 - 2 | 199 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |