Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342626.1 | NP_001331798.1 | 2 - 3 | 783 | 101 | |
NM_120270.4 | NP_195812.1 | 2 - 3 | 783 | 101 | ||
GenBank mRNA | AK117628.1 | - | 2 - 3 | 758 | 101 | |
BT006132.1 | - | 2 - 3 | 736 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |