Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P58IPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120394.4 | NP_195936.1 | 6 - 7 | 1147 | 75 | |
GenBank mRNA | AY085329.1 | - | 6 - 7 | 1100 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |