Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120476.5 | NP_196014.1 | 5 - 6 | 780 | 98 | |
GenBank mRNA | AY050999.1 | - | 5 - 6 | 676 | 98 | |
BT000976.1 | - | 5 - 6 | 612 | 98 | ||
Z21970.1 | - | 635 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |