Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HXK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084452.2 | NP_001077921.1 | 3 - 4 | 861 | 59 | |
NM_127544.3 | NP_179576.1 | 4 - 5 | 892 | 59 | ||
GenBank mRNA | U28215.1 | - | 4 - 5 | 816 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |