Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115612.3 | NP_191311.1 | 6 - 7 | 910 | 83 | |
NM_180384.1 | NP_850715.1 | 6 - 7 | 897 | 83 | ||
NM_202722.1 | NP_974451.1 | 6 - 7 | 897 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |