Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 20 - 21 | 3380 | 83 | |
NM_180432.2 | NP_850763.1 | 19 - 20 | 3212 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |