Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335796.1 | NP_001318266.1 | 4 - 5 | 477 | 109 | |
NM_001335797.1 | NP_001324040.1 | 3 - 4 | 638 | 109 | ||
NM_127796.3 | NP_565532.1 | 3 - 4 | 463 | 109 | ||
GenBank mRNA | AY048261.1 | - | 3 - 4 | 438 | 109 | |
AY072625.1 | - | 3 - 4 | 329 | 109 | ||
U76846.1 | - | 3 - 4 | 452 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |