Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127843.5 | NP_179863.1 | 8 - 9 | 1224 | 146 | |
GenBank mRNA | AF428346.1 | - | 8 - 9 | 1136 | 146 | |
AK227067.1 | - | 8 - 9 | 1134 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |