Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036852.4 | NP_001031929.4 | 2 - 3 | 915 | 74 | |
NM_001343803.1 | NP_001331260.1 | 2 - 3 | 799 | 74 | ||
NM_001343804.1 | NP_001331261.1 | 3 - 4 | 781 | 74 | ||
NM_122252.4 | NP_568432.1 | 2 - 3 | 953 | 74 | ||
NM_180734.3 | NP_851065.1 | 2 - 3 | 819 | 74 | ||
GenBank mRNA | AB046717.1 | - | 1 - 2 | 459 | 74 | |
AY139759.1 | - | 2 - 3 | 875 | 74 | ||
BT004544.1 | - | 1 - 2 | 459 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |