Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZO2H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344083.1 | NP_001330784.1 | 4 - 5 | 846 | 134 | |
NM_122760.4 | NP_568508.2 | 5 - 6 | 834 | 134 | ||
NM_180757.3 | NP_851088.1 | 5 - 6 | 745 | 134 | ||
GenBank mRNA | AF446876.1 | - | 5 - 6 | 655 | 134 | |
AK317169.1 | - | 5 - 6 | 690 | 134 | ||
AY052688.1 | - | 5 - 6 | 660 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |