Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 19 - 20 | 2010 | 69 | |
NM_124502.3 | NP_199936.2 | 20 - 21 | 2002 | 69 | ||
NM_180837.3 | NP_851168.1 | 20 - 21 | 2109 | 69 | ||
GenBank mRNA | AB053171.1 | - | 20 - 21 | 1954 | 69 | |
AY140086.1 | - | 20 - 21 | 1951 | 69 | ||
BT008388.1 | - | 19 - 20 | 1691 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |