Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203652.1 | NP_001190581.1 | 3 - 4 | 933 | 105 | |
NM_125457.3 | NP_568926.2 | 3 - 4 | 933 | 105 | ||
NM_180903.4 | NP_851234.1 | 3 - 4 | 993 | 105 | ||
GenBank mRNA | BT001994.1 | - | 3 - 4 | 931 | 105 | |
BT008359.1 | - | 3 - 4 | 888 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |