Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035762.6 | NP_001030839.5 | 4 - 5 | 3027 | 76 | |
NM_115036.7 | NP_190745.6 | 4 - 5 | 3021 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |