Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115171.3 | NP_190879.1 | 8 - 9 | 1560 | 101 | |
GenBank mRNA | BT002444.1 | - | 8 - 9 | 1511 | 101 | |
BT008867.1 | - | 7 - 8 | 1429 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |