Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084818.1 | NP_001078287.1 | 3 - 4 | 372 | 66 | |
NM_001339701.1 | NP_001327724.1 | 2 - 3 | 290 | 66 | ||
NM_115341.3 | NP_567008.1 | 3 - 4 | 372 | 66 | ||
GenBank mRNA | AB007766.2 | - | 3 - 4 | 282 | 66 | |
AK316975.1 | - | 3 - 4 | 323 | 66 | ||
BT002860.1 | - | 3 - 4 | 277 | 66 | ||
BT004392.1 | - | 3 - 4 | 187 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |