Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115359.3 | NP_191061.1 | 7 - 8 | 932 | 78 | |
NM_202708.2 | NP_974437.1 | 6 - 7 | 1049 | 78 | ||
GenBank mRNA | AY060585.1 | - | 7 - 8 | 910 | 78 | |
AY142054.1 | - | 6 - 7 | 836 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |