Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339744.1 | NP_001325527.1 | 4 - 5 | 1205 | 134 | |
NM_115432.5 | NP_191133.1 | 5 - 6 | 1083 | 134 | ||
NM_202715.2 | NP_974444.1 | 5 - 6 | 1067 | 134 | ||
GenBank mRNA | AY081292.1 | - | 5 - 6 | 904 | 134 | |
BT008750.1 | - | 5 - 6 | 730 | 134 | ||
X95738.1 | - | 881 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |