Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203183.1 | NP_001190112.1 | 3 - 4 | 554 | 94 | |
NM_115553.4 | NP_191253.1 | 2 - 3 | 631 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |