Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343870.1 | NP_001318635.1 | 2 - 3 | 411 | 104 | |
NM_001343871.1 | NP_001330711.1 | 2 - 3 | 432 | 104 | ||
NM_001343872.1 | NP_001330710.1 | 2 - 3 | 371 | 104 | ||
NM_122337.5 | NP_197819.2 | 2 - 3 | 447 | 104 | ||
GenBank mRNA | AK222217.1 | - | 2 - 3 | 369 | 104 | |
AK228814.1 | - | 2 - 3 | 249 | 104 | ||
AY817014.1 | - | 1 - 2 | 97 | 104 | ||
BT011734.1 | - | 1 - 2 | 97 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |