Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103537.5 | NP_175077.1 | 4 - 5 | 877 | 69 | |
GenBank mRNA | AK175406.1 | - | 4 - 5 | 772 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |