Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333320.1 | NP_001321802.1 | 3 - 4 | 593 | 76 | |
NM_103724.2 | NP_175261.1 | 3 - 4 | 593 | 76 | ||
GenBank mRNA | AK228479.1 | - | 3 - 4 | 446 | 76 | |
U95143.1 | - | 3 - 4 | 589 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |