Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115747.4 | NP_191444.1 | 1 | 889 | 158 | |
GenBank mRNA | AK229232.1 | - | 1 | 441 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |