Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203212.1 | NP_001190141.1 | 2 - 3 | 235 | 99 | |
NM_001340039.1 | NP_001327362.1 | 2 - 3 | 228 | 99 | ||
NM_001340040.1 | NP_001327363.1 | 1 - 2 | 262 | 99 | ||
NM_115897.5 | NP_191592.5 | 1 - 2 | 233 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |