Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128552.4 | NP_180559.1 | 4 - 5 | 806 | 94 | |
NM_179808.2 | NP_850139.1 | 4 - 5 | 653 | 94 | ||
GenBank mRNA | AY063966.1 | - | 4 - 5 | 668 | 94 | |
AY096462.1 | - | 4 - 5 | 562 | 94 | ||
D17579.1 | - | 4 - 5 | 644 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |