Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202708.1 | NP_001189637.1 | 7 - 8 | 1443 | 104 | |
NM_128582.2 | NP_180588.1 | 7 - 8 | 1446 | 104 | ||
GenBank mRNA | DQ446580.1 | - | 7 - 8 | 1312 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |