Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343268.1 | NP_001330075.1 | 4 - 5 | 1526 | 90 | |
NM_001343269.1 | NP_001330076.1 | 3 - 4 | 1539 | 90 | ||
NM_121335.4 | NP_196836.1 | 4 - 5 | 1316 | 90 | ||
GenBank mRNA | EU214910.1 | - | 3 - 4 | 1169 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |