Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343435.1 | NP_001331015.1 | 4 - 5 | 934 | 85 | |
NM_121610.4 | NP_568325.1 | 4 - 5 | 835 | 85 | ||
GenBank mRNA | AY035170.1 | - | 4 - 5 | 805 | 85 | |
AY059115.1 | - | 3 - 4 | 725 | 85 | ||
L09109.1 | - | 3 - 4 | 731 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |