Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336307.1 | NP_001324991.1 | 9 - 10 | 1117 | 116 | |
NM_128644.4 | NP_180649.1 | 8 - 9 | 967 | 116 | ||
GenBank mRNA | AY045828.1 | - | 8 - 9 | 958 | 116 | |
AY091366.1 | - | 8 - 9 | 850 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |