Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 25 - 26 | 3663 | 150 | |
NM_001336352.1 | NP_001323600.1 | 23 - 24 | 2950 | 150 | ||
NM_001336353.1 | NP_001323601.1 | 18 - 19 | 2176 | 150 | ||
NM_001336354.1 | NP_001323599.1 | 21 - 22 | 2741 | 150 | ||
NM_128748.3 | NP_180749.2 | 23 - 24 | 2950 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |